MilliporeSigma
Involvement of Golgin-160 in cell surface transport of renal ROMK channel: co-expression of Golgin-160 increases ROMK currents.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2006-03-18)
Florian Bundis, Ioana Neagoe, Blanche Schwappach, Klaus Steinmeyer
RÉSUMÉ
The weak inward rectifier potassium channel ROMK is important for water and salt reabsorption in the kidney. Here we identified Golgin-160 as a novel interacting partner of the ROMK channel. By using yeast two-hybrid assays and co-immunoprecipitations from transfected cells, we demonstrate that Golgin-160 associates with the ROMK C-terminus. Immunofluorescence microscopy confirmed that both proteins are co-localized in the Golgi region. The interaction was further confirmed by the enhancement of ROMK currents by the co-expressed Golgin-160 in Xenopus oocytes. The increase in ROMK current amplitude was due to an increase in cell surface density of ROMK protein. Golgin-160 also stimulated current amplitudes of the related Kir2.1, and of voltage-gated Kv1.5 and Kv4.3 channels, but not the current amplitude of co-expressed HERG channel. These results demonstrate that the Golgi-associated Golgin-160 recognizes the cytoplasmic C-terminus of ROMK, thereby facilitating the transport of ROMK to the cell surface. However, the stimulatory effect on the activity of more distantly-related potassium channels suggests a more general role of Golgin-160 in the trafficking of plasma membrane proteins.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Roche
Anti-Protein C Affinity Matrix, from mouse IgG1κ