MilliporeSigma
DOT1L activity in leukemia cells requires interaction with ubiquitylated H2B that promotes productive nucleosome binding.
Cell reports (2022-02-17)
Cathy J Spangler, Satya P Yadav, Dongxu Li, Carinne N Geil, Charlotte B Smith, Gang Greg Wang, Tae-Hee Lee, Robert K McGinty
RÉSUMÉ
DOT1L methylates histone H3 lysine 79 during transcriptional elongation and is stimulated by ubiquitylation of histone H2B lysine 120 (H2BK120ub) in a classical trans-histone crosstalk pathway. Aberrant genomic localization of DOT1L is implicated in mixed lineage leukemia (MLL)-rearranged leukemias, an aggressive subset of leukemias that lacks effective targeted treatments. Despite recent atomic structures of DOT1L in complex with H2BK120ub nucleosomes, fundamental questions remain as to how DOT1L-ubiquitin and DOT1L-nucleosome acidic patch interactions observed in these structures contribute to nucleosome binding and methylation by DOT1L. Here, we combine bulk and single-molecule biophysical measurements with cancer cell biology to show that ubiquitin and cofactor binding drive conformational changes to stimulate DOT1L activity. Using structure-guided mutations, we demonstrate that ubiquitin and nucleosome acidic patch binding by DOT1L are required for cell proliferation in the MV4; 11 leukemia model, providing proof of principle for MLL targeted therapeutic strategies.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys79) Antibody, clone NL59, rabbit monoclonal, culture supernatant, clone NL59, Upstate®