MilliporeSigma
[Juvenile hip pain. 2. Femur head epiphysiolysis, hip dysplasia, tumors].
Fortschritte der Medizin (1978-11-09)
W Hackenbruch, S von Gumppenberg, P M Karpf
PMID711104
RÉSUMÉ
The early symptom in hip joint diseases in children is pain. Pain is localized in the groin and thight, but mostly in the knee. Other important signs are limping and reduced internal rotation. If a hip disease is suspected it is necessary to take X-rays in two planes. If diagnosis is early and special therapy started immediately, the results are usually excellent without deformation of the hip. Otherwise early osteoarthritis can develop. This is important because osteoarthritis in the hip joint is in 75% of the cases due to hip joint diseases in childhood. The problems of diagnosis and treatment of the most common hip joint diseases in children (transient synovitis, rheumatoid arthritis, osteomyelitis, Legg-Perthes disease, slipped capital femoral epiphysis, dysplasia, tumors) are discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
1-Naphthylacetamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland