MilliporeSigma
  • Levels of endogenous β-glucanase activity in barley affect the efficacy of exogenous enzymes used to supplement barley-based diets for poultry.
Levels of endogenous β-glucanase activity in barley affect the efficacy of exogenous enzymes used to supplement barley-based diets for poultry.
Poultry science (2011-05-21)
T Ribeiro, M M S Lordelo, P I P Ponte, B Maçãs, J A M Prates, M Aguiar Fontes, L Falcão, J P B Freire, L M A Ferreira, C M G A Fontes
RÉSUMÉ
To improve the nutritive value of barley-based diet for broilers, 2 experiments using 2 different barley lots were performed to evaluate the capacity of a mesophilic cellulase when fused to a β-glucan specific family 11 carbohydrate-binding module. The data revealed that the recombinant β-glucanase derivatives were not appropriate for feed supplementation because of a lack of stability at acidic pH levels. However, under the same experimental conditions, a commercial enzyme mixture improved the nutritive value of 1 of the cereal lots used. Analysis of the nutritive value of the 2 barleys revealed intrinsic differences in the levels of endogenous β-glucanase activity. These differences were extensively evident when the studies were expanded to a range of 64 barley lots. Thus, to clarify the effect of endogenous cellulases on the efficacy of exogenous β-glucanases used to supplement barley-based diets for poultry, 2 barley lots presenting low and high levels of endogenous plant cell wall-degrading enzymes were selected. These lots were used to prepare 2 barley-based diets, which were supplemented with or without a commercial enzyme product and fed to broiler chicks. The data revealed that the exogenous enzymes were effective when the basal diet presented low levels of endogenous β-glucanases but were unable to improve the nutritive value of the barley lot displaying higher β-glucanase activity. Thus, these studies suggest that levels of endogenous β-glucanases may affect the efficacy of exogenous enzymes used to improve the nutritive value of barley-based diets for broilers. The development of a quick β-glucanase assay that could be applied for cereal-based feeds may help identify those barley-based diets that are more responsive to the action of feed enzymes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
β-Glucanase 2, thermostable, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)