MilliporeSigma
Role of CD38, a cyclic ADP-ribosylcyclase, in morphine antinociception and tolerance.
The Journal of pharmacology and experimental therapeutics (2010-06-17)
Lynn C Hull, Christopher Rabender, Bichoy H Gabra, Fan Zhang, Pin-Lan Li, William L Dewey
RÉSUMÉ
Our previous studies have demonstrated that an increase in intracellular levels of Ca(2+) in neurons is an important component of both the antinociception produced by morphine and morphine's tolerance. The present study tested the hypothesis that the Ca(2+) signaling second messenger, cyclic ADP-ribose (cADPR), derived from CD38 activation participates in morphine antinociception and tolerance. We first showed that morphine's antinociceptive potency was increased by the intracerebroventricular injection of CD38 substrate beta-NAD(+) in mice. Furthermore, morphine tolerance was reversed by intracerebroventricular administration of each of three different inhibitors of the CD38-cADPR-ryanodine receptor Ca(2+) signaling pathway. These inhibitors were the ADP-ribosylcyclase inhibitor nicotinamide, cADPR analog 8-bromo-cADPR, and a large dose of ryanodine (>50 muM) that blocks the ryanodine receptor. In CD38 gene knockout [CD38(-/-)] mice, the antinociceptive action of morphine was found to be less potent compared with wild-type (WT) mice, as measured by tail-flick response, hypothermia assay, and observations of straub tail. However, there was no difference in locomotor activation between CD38(-/-) and WT animals. It was also found that less tolerance to morphine developed in CD38(-/-) mice compared with WT animals. These results indicate that cADRP-ryanodine receptor Ca(2+) signaling associated with CD38 plays an important role in morphine tolerance.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
ADP-ribosylcyclase, 1000-3000 units/mg protein