MilliporeSigma
  • Dietary supplementation of a Saccharomyces cerevisiae fermentation product attenuates exercise-induced stress markers in young horses.
Dietary supplementation of a Saccharomyces cerevisiae fermentation product attenuates exercise-induced stress markers in young horses.
Journal of animal science (2021-06-29)
Hannah C Valigura, Jessica L Leatherwood, Rafael E Martinez, Sharon A Norton, Sarah H White-Springer
RÉSUMÉ
Mitigation of exercise-induced stress is of key interest in determining ways to optimize performance horse health. To test the hypothesis that dietary supplementation of a Saccharomyces cerevisiae fermentation product would decrease markers of exercise-induced stress and inflammation in young horses, Quarter Horse yearlings (mean ± SD; 9 ± 1 mo) were randomly assigned to receive either no supplementation (CON; n = 8) or 21 g/d S. cerevisiae fermentation product (10.5 g/feeding twice daily; SCFP; n = 10) top-dressed on a basal diet of custom-formulated grain as well as ad libitum Coastal bermudagrass hay. After 8 wk of dietary treatments, horses underwent a 2-h submaximal exercise test (SET) on a free-stall mechanical exerciser. Serum was collected before dietary treatment supplementation (week 0), at week 8 pre-SET, and 0, 1, and 6 h post-SET and analyzed for concentrations of cortisol and serum amyloid A (SAA) by commercial enzyme-linked immunosorbent assay (ELISA) and for cytokine concentrations by commercial bead-based ELISA. Data were analyzed using linear models with repeated measures in SAS v9.4. From week 0 to 8 (pre-SET), serum cortisol decreased (P = 0.01) and SAA did not change, but neither were affected by diet. Serum concentrations of all cytokines decreased from week 0 to 8 (P ≤ 0.008), but granulocyte colony-stimulating factor (G-CSF), granulocyte-macrophage colony-stimulating factor, and interleukin-8 (IL-8) decreased to a greater extent in CON than in SCFP horses (P ≤0.003). In response to the week 8 SET, serum cortisol increased in all horses (P < 0.0001) but returned to pre-SET levels by 1 h post-SET in horses receiving SCFP. At 6 h post-SET, cortisol concentrations in CON horses returned to pre-SET concentrations, whereas cortisol declined further in SCFP horses to below pre-SET levels (P = 0.0002) and lower than CON (P = 0.003) at that time point. SAA increased at 6 h post-SET in CON (P < 0.0001) but was unchanged through 6 h in SCFP horses. All cytokines except G-CSF increased in response to the SET (P < 0.0001) but showed differing response patterns. Concentrations of IL-1β, IL-6, and tumor necrosis factor-alpha were lesser (P ≤ 0.05), and concentrations of G-CSF and IL-18 tended to be lesser (P ≤ 0.09) in SCFP compared with CON horses throughout recovery from the SET. In summary, 8 wk of dietary supplementation with 21 g/d of SCFP may mitigate cellular stress following a single, prolonged submaximal exercise bout in young horses.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Millipore
Panel de cytokines/chimiokines équines MILLIPLEX® avec billes magnétiques - 23 plex prémixés - Essai multiplex d'immunologie, The Equine Inflammation / Immunology Bead-Based Multiplex Cytokine Assay kit, using the Luminex xMAP technology, enables the simultaneous analysis of 23 cytokine and chemokine biomarkers in equine serum, plasma and cell culture samples.