MilliporeSigma
  • Cortexolone 17α-propionate (Clascoterone) Is a Novel Androgen Receptor Antagonist that Inhibits Production of Lipids and Inflammatory Cytokines from Sebocytes In Vitro
Cortexolone 17α-propionate (Clascoterone) Is a Novel Androgen Receptor Antagonist that Inhibits Production of Lipids and Inflammatory Cytokines from Sebocytes In Vitro
Journal of drugs in dermatology : JDD (2019-05-31)
Caridad Rosette, Frances J. Agan, Alessandro Mazzetti, Luigi Moro, Mara Gerloni
RÉSUMÉ
Cortexolone 17α-propionate (clascoterone) is a novel topical androgen antagonist that is being analyzed for its ability to treat acne. The pathogenesis of acne is attributed to multiple factors, including altered sebum production, inflammatory processes, dysregulation of the hormone microenvironment, and the proliferation of the skin commensal bacteria, Propionibacterium acnes (P. acnes). Androgens induce the proliferation and differentiation of sebocytes, (cells that comprise the sebaceous gland), help regulate the synthesis of the lipids that are incorporated into sebum and stimulate the production of cytokines that are found in inflammatory acne lesions. Several studies have established that clascoterone is a potent antiandrogen that is well tolerated and has selective topical activity. Its potency as an acne therapeutic is currently being analyzed in a large phase 3 clinical trial. The study described herein elucidates for the first time the mechanism of action of clascoterone. Clascoterone was found to bind the androgen receptor (AR) with high affinity in vitro, inhibit AR-regulated transcription in a reporter cell line, and antagonize androgen-regulated lipid and inflammatory cytokine production in a dose-dependent manner in human primary sebocytes. In particular, when compared to another AR antagonist, spironolactone, clascoterone was significantly better at inhibiting inflammatory cytokine synthesis from sebocytes. Therefore, clascoterone may be an excellent candidate to be the first topical antiandrogen to treat acne. J Drugs Dermatol. 2019;18(5):412-418.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Clascoterone, ≥98% (HPLC)