MilliporeSigma
The Origin, Expression, Function and Future Research Focus of a G Protein-coupled Receptor, Mas-related Gene X2 (MrgX2).
Progress in histochemistry and cytochemistry (2015-06-25)
Haihui Wu, Meiqi Zeng, Eric Y P Cho, Wenqi Jiang, Ou Sha
RÉSUMÉ
Mas-related genes (Mrgs) belong to a large family of G protein-coupled receptor genes found in rodents. Human MRGX proteins are G protein-coupled 7-transmembrane proteins sharing 41-52% amino acid identity with each other, but have no orthologs in rodents. MrgX2 is a member of the MrgX family. MRGX2 is expressed in the small neurons of sensory ganglia and mast cells. It can interact with a series of factors and genes such as the peptides substance P, vasoactive intestinal peptide, cortistatin (CST), proadrenomedullin N-terminal peptide (PAMP), LL-37, PMX-53 and β-defensins. MRGX2 is related to nociception, adrenal gland secretion and mast cell degranulation. Recent research on MrgX2 provides insights into its role in nociception and anti-microbial activities. This article reviewed the origin, expression and function of MrgX2, and discussed possible future research focus.