MilliporeSigma
  • Upregulation of C/EBPα Inhibits Suppressive Activity of Myeloid Cells and Potentiates Antitumor Response in Mice and Patients with Cancer.
Upregulation of C/EBPα Inhibits Suppressive Activity of Myeloid Cells and Potentiates Antitumor Response in Mice and Patients with Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2021-08-20)
Ayumi Hashimoto, Debashis Sarker, Vikash Reebye, Sheba Jarvis, Mikael H Sodergren, Andrew Kossenkov, Emilio Sanseviero, Nina Raulf, Jenni Vasara, Pinelopi Andrikakou, Tim Meyer, Kai-Wen Huang, Ruth Plummer, Cheng E Chee, Duncan Spalding, Madhava Pai, Shahid Khan, David J Pinato, Rohini Sharma, Bristi Basu, Daniel Palmer, Yuk-Ting Ma, Jeff Evans, Robert Habib, Anna Martirosyan, Naouel Elasri, Adeline Reynaud, John J Rossi, Mark Cobbold, Nagy A Habib, Dmitry I Gabrilovich
RÉSUMÉ
To evaluate the mechanisms of how therapeutic upregulation of the transcription factor, CCAAT/enhancer-binding protein alpha (C/EBPα), prevents tumor progression in patients with advanced hepatocellular carcinoma (HCC) and in different mouse tumor models. We conducted a phase I trial in 36 patients with HCC (NCT02716012) who received sorafenib as part of their standard care, and were given therapeutic C/EBPα small activating RNA (saRNA; MTL-CEBPA) as either neoadjuvant or adjuvant treatment. In the preclinical setting, the effects of MTL-CEBPA were assessed in several mouse models, including BNL-1ME liver cancer, Lewis lung carcinoma (LLC), and colon adenocarcinoma (MC38). MTL-CEBPA treatment caused radiologic regression of tumors in 26.7% of HCC patients with an underlying viral etiology with 3 complete responders. MTL-CEBPA treatment in those patients caused a marked decrease in peripheral blood monocytic myeloid-derived suppressor cell (M-MDSC) numbers and an overall reduction in the numbers of protumoral M2 tumor-associated macrophages (TAM). Gene and protein analysis of patient leukocytes following treatment showed CEBPA activation affected regulation of factors involved in immune-suppressive activity. To corroborate this observation, treatment of all the mouse tumor models with MTL-CEBPA led to a reversal in the suppressive activity of M-MDSCs and TAMs, but not polymorphonuclear MDSCs (PMN-MDSC). The antitumor effects of MTL-CEBPA in these tumor models showed dependency on T cells. This was accentuated when MTL-CEBPA was combined with checkpoint inhibitors or with PMN-MDSC-targeted immunotherapy. This report demonstrates that therapeutic upregulation of the transcription factor C/EBPα causes inactivation of immune-suppressive myeloid cells with potent antitumor responses across different tumor models and in cancer patients. MTL-CEBPA is currently being investigated in combination with pembrolizumab in a phase I/Ib multicenter clinical study (NCT04105335).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-LOX-1, clone 9E12.1, clone 9E12.1, from mouse