MilliporeSigma
Synthesis of novel acetylenic cyclophanes with helical chirality: potential new structures for liquid crystals
Organic letters (2000-09-29)
Collins, Yap, Fallis
RÉSUMÉ
The synthesis of a series of novel acetylenic cyclophanes is described. X-ray crystallographic analysis of the core structure revealed a twisted conformation with helical chirality. Preliminary results suggest that these cyclophanes, with appropriate functionality, have the potential to act as unique liquid crystalline materials.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Methyl 11-bromoundecanoate, 95%