MilliporeSigma
Uncovering an allosteric mode of action for a selective inhibitor of human Bloom syndrome protein.
eLife (2021-03-02)
Xiangrong Chen, Yusuf I Ali, Charlotte El Fisher, Raquel Arribas-Bosacoma, Mohan B Rajasekaran, Gareth Williams, Sarah Walker, Jessica R Booth, Jessica Jr Hudson, S Mark Roe, Laurence H Pearl, Simon E Ward, Frances Mg Pearl, Antony W Oliver
RÉSUMÉ
BLM (Bloom syndrome protein) is a RECQ-family helicase involved in the dissolution of complex DNA structures and repair intermediates. Synthetic lethality analysis implicates BLM as a promising target in a range of cancers with defects in the DNA damage response; however, selective small molecule inhibitors of defined mechanism are currently lacking. Here, we identify and characterise a specific inhibitor of BLM's ATPase-coupled DNA helicase activity, by allosteric trapping of a DNA-bound translocation intermediate. Crystallographic structures of BLM-DNA-ADP-inhibitor complexes identify a hitherto unknown interdomain interface, whose opening and closing are integral to translocation of ssDNA, and which provides a highly selective pocket for drug discovery. Comparison with structures of other RECQ helicases provides a model for branch migration of Holliday junctions by BLM.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
ML216, ≥98% (HPLC)