MilliporeSigma
Expression of the TrkB neurotrophin receptor by thymic macrophages.
Immunology (1998-09-19)
O García-Suárez, J Hannestad, I Esteban, R Sainz, F J Naves, J A Vega
RÉSUMÉ
Increasing evidence suggests that some members of the neurotrophic factor family of neurotrophins could be implicated in the regulation of immune responses. Neurotrophins, as well as their tyrosine kinase signal-transducing receptors (the so-called Trk neurotrophin receptors), have been detected in different lymphoid tissues, although their cellular localization is not well known. In this study we used single and double immunohistochemistry to localize TrkB in situ in the rat thymus (in animals from 0 days to 2 years of age), in cytospin preparations of rat thymic cells, and in two mouse monocyte-macrophage cell lines (RAW 264.7 and J774A.1). We found TrkB protein expression in a subpopulation of cells in the corticomedullary junction, which simultaneously expressed the rat macrophage marker ED1. The density of TrkB-expressing cells increased with age, reaching maximal values at 2 years. Conversely, no evidence of TrkB protein expression could be found in dendritic cells, epithelial cells or thymocytes. Thymic macrophages in cytospin preparations, as well as in the mouse monocyte macrophage cell lines, also expressed TrkB protein. Although the possible function of TrkB in the thymic macrophage remains to be clarified, present findings add further evidence to the proposed role of neurotrophins in the immune system.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Boc-Ser-OH, ≥99.0% (T)