MilliporeSigma
An ezrin/calpain/PI3K/AMPK/eNOSs1179 signaling cascade mediating VEGF-dependent endothelial nitric oxide production.
Circulation research (2008-11-29)
Ji-Youn Youn, Ting Wang, Hua Cai
RÉSUMÉ
Calpain was recently reported to mediate vascular endothelial growth factor (VEGF)-induced angiogenesis. In the present study, we investigated detailed molecular mechanisms. VEGF (100 ng/mL) induced a marked increase in endothelial cell production of NO(*), specifically detected by electron spin resonance. This response was abolished by inhibition of calpain with N-acetyl-leucyl-leucyl-norleucinal (ALLN) or Calpeptin. Both also diminished membrane-specific calpain activation by VEGF, which was intriguingly attenuated by silencing ezrin with RNA interference. A rapid membrane colocalization of calpain and ezrin occurred as short as 10 minutes after VEGF stimulation. AKT, AMP-dependent kinase (AMPK), and endothelial nitric oxide synthase (eNOS)(s1179) phosphorylations in VEGF-stimulated endothelial cells were markedly enhanced, which were however significantly attenuated by either ALLN, Calpeptin, or ezrin small interfering RNA, as well as by Wortmannin or compound C (respectively for phosphatidylinositol 3-kinase [PI3K] or AMPK). The latter 3 also abolished VEGF induction of NO(*). These data indicate that AMPK and AKT are both downstream of PI3K and that AKT activation is partially dependent on AMPK. The interrelationship between AMPK and AKT, although known to be individually important in mediating VEGF activation of eNOS, is clearly characterized. Furthermore, AMPK/AKT/eNOS(s1179) was found downstream of a calpain/ezrin membrane interaction. These data no doubt provide new insights into the long mystified signaling gap between VEGF receptors and PI3K/AKT or AMPK-dependent eNOS activation. In view of the well-established significance of VEGF-dependent angiogenesis, these findings might have broad and important implications in cardiovascular pathophysiology.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Calpain Activity Assay Kit, Fluorogenic