MilliporeSigma
Monitoring of serum and urinary biomarkers during treatment of canine visceral leishmaniasis.
Veterinary world (2020-10-17)
Alvaro Felipe de Lima Ruy Dias, Eveline da Cruz Boa Sorte Ayres, Fernanda Harumi Maruyama, Bruna Ribeiro Gomes Monteiro, Maria Sabrina de Freitas, Arleana do Bom Parto Ferreira de Almeida, Adriane Jorge Mendonça, Valéria Régia Franco Sousa
RÉSUMÉ
Canine visceral leishmaniasis (CanL) has a broad spectrum of changes, with kidney disease being considered the main cause of mortality. Thus, this study aimed to monitor serum and urinary biomarkers in response to two short-term treatments for CanL. Thirty dogs with CanL were equally divided into two treatment groups and treated with either miltefosine (Group M) or miltefosine plus allopurinol (Group MA); the groups were evaluated before treatment and after 28 days of treatment. Physical exams were performed and hematimetric, biochemical, and urinary parameters, including urinary biomarkers cystatin C (CisC), lipocalin-2 (NGAL), and microalbuminuria, were measured. Both treatments significantly reduced clinical scores (p<0.05), but only the MA group saw a reduction in the clinical-pathological score. The serum albumin and calcium levels increased significantly in the MA and M groups (p<0.05). Proteinuria and urinary density did not decrease significantly after the treatments. With regard to the biomarkers, CisC and microalbuminuria did not have any significant changes; however, NGAL was significantly reduced in the MA group (p<0.05). Both pharmacotherapeutic protocols promoted clinical and clinical-pathological improvements. In addition, miltefosine plus allopurinol proved to be a safe treatment due to the lack of changes detected in the monitored renal biomarkers. The treatment with miltefosine plus allopurinol proved to be the most effective, with more pronounced beneficial effects for canines with visceral leishmaniasis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Millipore
MILLIPLEX® Canine Kidney Toxicity Expanded Magnetic Bead Panel 1, CKT1MAG-97K, The analytes available for this multiplex kit are: Clusterin, Cystatin C, Kidney Injury Molecule-1 (KIM-1), IL-8, Lipocalin-2/NGAL, MCP-1, Osteopontin (OPN).