- Perennisosides I-VII, acylated triterpene saponins with antihyperlipidemic activities from the flowers of Bellis perennis.
Perennisosides I-VII, acylated triterpene saponins with antihyperlipidemic activities from the flowers of Bellis perennis.
Journal of natural products (2008-03-28)
Toshio Morikawa, Xuezheng Li, Eriko Nishida, Yuki Ito, Hisashi Matsuda, Seikou Nakamura, Osamu Muraoka, Masayuki Yoshikawa
PMID18363378
RÉSUMÉ
The methanolic extract and its saponin fraction (methanol-eluted fraction) of the flowers of Bellis perennis were found to suppress serum triglyceride elevation in olive oil-treated mice. From the saponin fraction, seven new triterpene saponins, perennisosides I (1), II (2), III (3), IV (4), V (5), VI (6), and VII (7), were isolated together with four known saponins, bellidioside A (8), asterbatanoside D (9), bernardioside B 2 (10), and bellissaponin BS6 (11). The stereostructures of 1- 7 were elucidated on the basis of chemical and spectroscopic evidence. Among these saponins, perennisosides I (1) and II (2) showed inhibitory effects on serum triglyceride elevation at doses of 25-50 mg/kg, po.
MATÉRIAUX