MilliporeSigma
  • Intermittent hypoxia degrades HIF-2alpha via calpains resulting in oxidative stress: implications for recurrent apnea-induced morbidities.
Intermittent hypoxia degrades HIF-2alpha via calpains resulting in oxidative stress: implications for recurrent apnea-induced morbidities.
Proceedings of the National Academy of Sciences of the United States of America (2009-01-17)
Jayasri Nanduri, Ning Wang, Guoxiang Yuan, Shakil A Khan, Dangjai Souvannakitti, Ying-Jie Peng, Ganesh K Kumar, Joseph A Garcia, Nanduri R Prabhakar
RÉSUMÉ
Intermittent hypoxia (IH) occurs in many pathological conditions including recurrent apneas. Hypoxia-inducible factors (HIFs) 1 and 2 mediate transcriptional responses to low O(2). A previous study showed that HIF-1 mediates some of the IH-evoked physiological responses. Because HIF-2alpha is an orthologue of HIF-1alpha, we examined the effects of IH on HIF-2alpha, the O(2)-regulated subunit expression, in pheochromocytoma 12 cell cultures. In contrast to the up-regulation of HIF-1alpha, HIF-2alpha was down-regulated by IH. Similar down-regulation of HIF-2alpha was also seen in carotid bodies and adrenal medullae from IH-exposed rats. Inhibitors of calpain proteases (ALLM, ALLN) prevented IH-evoked degradation of HIF-2alpha whereas inhibitors of prolyl hydroxylases or proteosome were ineffective. IH activated calpain proteases and down-regulated the endogenous calpain inhibitor calpastatin. IH-evoked HIF-2alpha degradation led to inhibition of SOD2 transcription, resulting in oxidative stress. Over-expression of transcriptionally active HIF-2alpha prevented IH-evoked oxidative stress and restored SOD2 activity. Systemic treatment of IH-exposed rats with ALLM rescued HIF-2alpha degradation and restored SOD2 activity, thereby preventing oxidative stress and hypertension. These observations demonstrate that, unlike continuous hypoxia, IH leads to down-regulation of HIF-2alpha via a calpain-dependent signaling pathway and results in oxidative stress as well as autonomic morbidities.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
InnoZyme Calpain 1/2 Activity Assay Kit