MilliporeSigma
Extensive Proliferation of Human Cancer Cells with Ever-Shorter Telomeres.
Cell reports (2017-06-22)
Rebecca A Dagg, Hilda A Pickett, Axel A Neumann, Christine E Napier, Jeremy D Henson, Erdahl T Teber, Jonathan W Arthur, C Patrick Reynolds, Jayne Murray, Michelle Haber, Alexander P Sobinoff, Loretta M S Lau, Roger R Reddel
RÉSUMÉ
Acquisition of replicative immortality is currently regarded as essential for malignant transformation. This is achieved by activating a telomere lengthening mechanism (TLM), either telomerase or alternative lengthening of telomeres, to counter normal telomere attrition. However, a substantial proportion of some cancer types, including glioblastomas, liposarcomas, retinoblastomas, and osteosarcomas, are reportedly TLM-negative. As serial samples of human tumors cannot usually be obtained to monitor telomere length changes, it has previously been impossible to determine whether tumors are truly TLM-deficient, there is a previously unrecognized TLM, or the assay results are false-negative. Here, we show that a subset of high-risk neuroblastomas (with ∼50% 5-year mortality) lacked significant TLM activity. Cancer cells derived from these highly aggressive tumors initially had long telomeres and proliferated for >200 population doublings with ever-shorter telomeres. This indicates that prevention of telomere shortening is not always required for oncogenesis, which has implications for inhibiting TLMs for cancer therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®