MilliporeSigma
Seleno-l-methionine suppresses copper-enhanced zinc-induced neuronal cell death via induction of glutathione peroxidase.
Metallomics : integrated biometal science (2020-09-15)
Yukari Nakano, Mikako Shimoda, Saki Okudomi, Sayuri Kawaraya, Masahiro Kawahara, Ken-Ichiro Tanaka
RÉSUMÉ
Excessive zinc ion (Zn2+) release is induced in pathological situations and causes neuronal cell death. Previously, we have reported that copper ions (Cu2+) markedly exacerbated Zn2+-induced neuronal cell death by potentiating oxidative stress, the endoplasmic reticulum (ER) stress response, and the activation of the c-Jun amino-terminal kinase (JNK) signaling pathway. In contrast, selenium (Se), an essential trace element, and amino acids containing selenium (such as seleno-l-methionine) have been reported to inhibit stress-induced neuronal cell death and oxidative stress. Thus, we investigated the effect of seleno-l-methionine on Cu2+/Zn2+-induced neuronal cell death in GT1-7 cells. Seleno-l-methionine treatment clearly restored the Cu2+/Zn2+-induced decrease in the viable cell number and attenuated the Cu2+/Zn2+-induced cytotoxicity. Accordingly, the levels of ER stress-related factors (especially, CHOP and GADD34) and of phosphorylated JNK increased upon CuCl2 and ZnCl2 co-treatment, whereas pre-treatment with seleno-l-methionine significantly suppressed these upregulations. Analysis of reactive oxygen species (ROS) as upstream factors of these pathways revealed that Cu2+/Zn2+-induced ROS production was clearly suppressed by seleno-l-methionine treatment. Finally, we found that seleno-l-methionine induced the antioxidative protein, glutathione peroxidase. Taken together, our findings suggest that seleno-l-methionine suppresses Cu2+/Zn2+-induced neuronal cell death and oxidative stress via induction of glutathione peroxidase. Thus, we think that seleno-l-methionine may help prevent refractory neurological diseases.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Seleno-L-methionine, ≥98% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human GPX1