MilliporeSigma
Engineered erythrocytes covalently linked to antigenic peptides can protect against autoimmune disease.
Proceedings of the National Academy of Sciences of the United States of America (2017-03-09)
Novalia Pishesha, Angelina M Bilate, Marsha C Wibowo, Nai-Jia Huang, Zeyang Li, Rhogerry Deshycka, Djenet Bousbaine, Hojun Li, Heide C Patterson, Stephanie K Dougan, Takeshi Maruyama, Harvey F Lodish, Hidde L Ploegh
RÉSUMÉ
Current therapies for autoimmune diseases rely on traditional immunosuppressive medications that expose patients to an increased risk of opportunistic infections and other complications. Immunoregulatory interventions that act prophylactically or therapeutically to induce antigen-specific tolerance might overcome these obstacles. Here we use the transpeptidase sortase to covalently attach disease-associated autoantigens to genetically engineered and to unmodified red blood cells as a means of inducing antigen-specific tolerance. This approach blunts the contribution to immunity of major subsets of immune effector cells (B cells, CD4+ and CD8+ T cells) in an antigen-specific manner. Transfusion of red blood cells expressing self-antigen epitopes can alleviate and even prevent signs of disease in experimental autoimmune encephalomyelitis, as well as maintain normoglycemia in a mouse model of type 1 diabetes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Acide polyinosinique–polycytidylique sodium salt, γ-irradiated
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, Alkaline Phosphatase conjugate, 0.6 mg/mL, Chemicon®