MilliporeSigma
In flow metal-enhanced fluorescence for biolabelling and biodetection.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2020-07-18)
Daniela Gontero, Alicia V Veglia, A Guillermo Bracamonte
RÉSUMÉ
Escherichia coli bacteria were determined by in flow cytometry with laser excitation and fluorescence detection applying ultraluminescent core-shell nanoparticles based on Metal Enhanced Fluorescence (MEF). Core-shell nanoparticles consisted of a 40 nm core modified with a silica spacer grafted with Rhodamine B (RhB). The electromagnetic field in the near field of the core surface enhanced the fluorescence of RhB by plasmonic and fluorophore coupling. The hydrophilic silica spacer allowed the non-covalent interaction with the polar E. coli surface and thus ultraluminescent bacteria biolabelling was developed. Clearly, well defined and bright bacteria imaging was recorded by Laser Fluorescence Microscopy based on the non-covalent deposition of the ultraluminescent nano-emitters. Using these nano-labellers, it was possible to detect labelled E. coli by in flow cytometry. Higher values of Side-scattered light (SSC) and Forward-scattered light (FSC), and number of fluorescent event detections, were observed for labelled bacteria compared to those non-labelled. The sensitivity of the methodology was evaluated by varying bacteria concentration and acceptable analytical figures of merit were determined. Applying this methodology we could quantify E. coli from a synthetic real sample of fortified water. Similar results were obtained by bacteria counting with Laser Fluorescence Microscopy and with a cell-bacteria counter.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
Sodium cyanide, ACS reagent, ≥95.0%