MilliporeSigma
  • Placental transfer of Letermovir & Maribavir in the ex vivo human cotyledon perfusion model. New perspectives for in utero treatment of congenital cytomegalovirus infection.
Placental transfer of Letermovir & Maribavir in the ex vivo human cotyledon perfusion model. New perspectives for in utero treatment of congenital cytomegalovirus infection.
PloS one (2020-05-01)
Valentine Faure Bardon, Gilles Peytavin, Minh Patrick Lê, Tiffany Guilleminot, Elisabeth Elefant, Julien Stirnemann, Marianne Leruez-Ville, Yves Ville
RÉSUMÉ
Congenital cytomegalovirus infection can lead to severe sequelae. When fetal infection is confirmed, we hypothesize that fetal treatment could improve the outcome. Maternal oral administration of an effective drug crossing the placenta could allow fetal treatment. Letermovir (LMV) and Maribavir (MBV) are new CMV antivirals, and potential candidates for fetal treatment. The objective was to investigate the placental transfer of LMV and MBV in the ex vivo method of the human perfused cotyledon. Term placentas were perfused, in an open-circuit model, with LMV or MBV at concentrations in the range of clinical peak plasma concentrations. Concentrations were measured using ultraperformance liquid chromatography coupled with tandem mass spectrometry. Mean fetal transfer rate (FTR) (fetal (FC) /maternal concentration), clearance index (CLI), accumulation index (AI) (retention of each drug in the cotyledon tissue) were measured. Mean FC were compared with half maximal effective concentrations of the drugs (EC50(LMV) and EC50(MBV)). For LMV, the mean FC was (± standard deviation) 1.1 ± 0.2 mg/L, 1,000-fold above the EC50(LMV). Mean FTR, CLI and AI were 9 ± 1%, 35 ± 6% and 4 ± 2% respectively. For MBV, the mean FC was 1.4 ± 0.2 mg/L, 28-fold above the EC50(MBV). Mean FTR, CLI and AI were 10 ± 1%, 50 ± 7% and 2 ± 1% respectively. Drugs' concentrations in the fetal side should be in the range for in utero treatment of fetuses infected with CMV as the mean FC was superior to the EC50 for both molecules.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Maribavir, ≥98% (HPLC)