MilliporeSigma
FRMD7 Mutations Disrupt the Interaction with GABRA2 and May Result in Infantile Nystagmus Syndrome.
Investigative ophthalmology & visual science (2020-05-24)
Lei Jiang, Yulei Li, Kangjuan Yang, Yuping Wang, Jiuxiang Wang, Xiaoniu Cui, Jinglin Mao, Yong Gao, Ping Yi, Lejin Wang, Jing Yu Liu
RÉSUMÉ
To identify the pathogenic gene of infantile nystagmus syndrome (INS) in three Chinese families and explore the potential pathogenic mechanism of FERM domain-containing 7 (FRMD7) mutations. Genetic testing was performed via Sanger sequencing. Western blotting was used to analyze protein expression of FRMD7. Glutathione S-transferase pull-down and immunoprecipitation were conducted to investigate the proteins interacting with FRMD7. Rescue assays were performed in Caenorhabditis elegans to explore the potential role of FRMD7 in vivo. We recruited three Chinese families with X-linked INS and identified a duplication and two missense mutations in FRMD7: c.998dupA/p.His333Glnfs*2, c.580G>A/p.Ala194Thr, and c.973A>G/p.Arg325Gly (one in each family). Expression levels of three mutants were similar to that of wild-type FRMD7 in vitro. Interestingly, the mutant p.His333Glnfs*2 exhibited a predominantly nuclear location, whereas wild-type FRMD7 localized to the cytoplasm. In addition, we found FRMD7 to directly interact with the loop between transmembrane domains 3 and 4 of GABRA2, a type A gamma-aminobutyric acid (GABA) receptor (GABAARs) subunit critical for receptor transport and localization, whereas the mutants p.Ala194Thr and p.Arg325Gly exhibited decreased binding to GABRA2. In frm-3 (a nematode homologue of FRMD7) null C. elegans, we found that FRMD7 mutants exhibited a poor rescue effect on the defects of locomotion and fluorescence recovery after photobleaching of GABAARs. Our findings identified three FRMD7 mutants in three Chinese families with X-linked INS and confirmed GABRA2 as a novel binding partner of FRMD7. These findings suggest that FRMD7 plays an important role by targeting GABAARs.