MilliporeSigma
  • Mesenchymal stromal cell-derived extracellular vesicles modulate microglia/macrophage polarization and protect the brain against hypoxia-ischemic injury in neonatal mice by targeting delivery of miR-21a-5p.
Mesenchymal stromal cell-derived extracellular vesicles modulate microglia/macrophage polarization and protect the brain against hypoxia-ischemic injury in neonatal mice by targeting delivery of miR-21a-5p.
Acta biomaterialia (2020-07-04)
Danqing Xin, Tingting Li, Xili Chu, Hongfei Ke, Zhuoya Yu, Lili Cao, Xuemei Bai, Dexiang Liu, Zhen Wang
RÉSUMÉ
Mesenchymal stromal cell (MSC)-derived extracellular vesicles (EVs) (MSC-EVs) exhibit protective effects in damaged or diseased tissues. However, the role of EVs secreted by MSC in hypoxia-ischemic (HI) injury in neonatal mice remains unknown. Systemic administration of MSC-EVs attenuated acute brain damage and neuroinflammation, and skewed CD11b+/CD45low microglia and CD11b+/CD45high brain monocyte/macrophage towards a more anti-inflammatory property as determined at 72 h post-HI. In addition, MSC-EVs remarkably improve the injury outcomes pups prior to weaning (P21), while no effect on long-term memory impairment (P42). Importantly, these effects were preceded by incorporation of MSC-EVs into a large number of neurons and microglia within HI group. Abundant levels of miR-21a-5p were present in EVs as determined with next-generation sequencing. Notably, MSC-EVs treatment further increased miR-21a-5p levels at 72 h post HI. Knockdown analyses revealed that miR-21a-5p, and its target-Timp3, were essential for this neuroprotective property of MSC-EVs following HI exposure as demonstrated in both in vitro and in vivo models. These findings suggest that a systemic administration of EVs derived from MSC, have the capacity to incorporated into neurons and microglia where they can then exert neuroprotection against HI-induced injury in neonates through the delivery of miR-21a-5p.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Timp3