MilliporeSigma
Glucagon-like peptide 1 receptor agonist ZP10A increases insulin mRNA expression and prevents diabetic progression in db/db mice.
The Journal of pharmacology and experimental therapeutics (2003-09-17)
Christian Thorkildsen, Søren Neve, Bjarne Due Larsen, Eddi Meier, Jørgen Søberg Petersen
RÉSUMÉ
We characterized the novel, rationally designed peptide glucagon-like peptide 1 (GLP-1) receptor agonist H-HGEGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPSK KKKKK-NH2 (ZP10A). Receptor binding studies demonstrated that the affinity of ZP10A for the human GLP-1 receptor was 4-fold greater than the affinity of GLP-1 (7-36) amide. ZP10A demonstrated dose-dependent improvement of glucose tolerance with an ED50 value of 0.02 nmol/kg i.p. in an oral glucose tolerance test (OGTT) in diabetic db/db mice. After 42 days of treatment, ZP10A dose-dependently (0, 1, 10, or 100 nmol/kg b.i.d.; n = 10/group), decreased glycosylated hemoglobin (HbA1C) from 8.4 +/- 0.4% (vehicle) to a minimum of 6.2 +/- 0.3% (100 nmol/kg b.i.d.; p < 0.05 versus vehicle) in db/db mice. Fasting blood glucose (FBG), glucose tolerance after an OGTT, and HbA1C levels were significantly improved in mice treated with ZP10A for 90 days compared with vehicle-treated controls. Interestingly, these effects were preserved 40 days after drug cessation in db/db mice treated with ZP10A only during the first 50 days of the study. Real-time polymerase chain reaction measurements demonstrated that the antidiabetic effect of early therapy with ZP10A was associated with an increased pancreatic insulin mRNA expression relative to vehicle-treated mice. In conclusion, long-term treatment of diabetic db/db mice with ZP10A resulted in a dose-dependent improvement of FBG, glucose tolerance, and blood glucose control. Our data suggest that ZP10A preserves beta-cell function. ZP10A is considered one of the most promising new drug candidates for preventive and therapeutic intervention in type 2 diabetes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Lixisenatide, ≥95% (HPLC)