- Analysis of arylamine isomers by micellar electrokinetic chromatography.
Analysis of arylamine isomers by micellar electrokinetic chromatography.
Electrophoresis (2002-03-01)
Rama Jeevan Ganesh Jeevan, Malayappan Bhaskar, Raghavan Chandrasekar, Ganga Radhakrishnan
PMID11870769
RÉSUMÉ
Analysis of carcinogenic substances is a high-priority area. Carcinogenic arylamines draw the analyst's attention because dyes and pigments are in production and used in large volumes. Identification of carcinogenic isomers of arylamines employing micellar electrokinetic chromatography (MEKC), a mode of capillary electrophoresis was studied as it offers better scope for separation science. Mixed micellar modes of MEKC techniques were employed to achieve acceptable analyses. Success of this analytical method was proved by real-sample analysis, which confirmed that this is a promising technique for the arylamine species.