MilliporeSigma
  • Insights into the Photodecomposition of Azidomethyl Methyl Sulfide: A S2/S1 Conical Intersection on Nitrene Potential Energy Surfaces Leading to the Formation of S-Methyl-N-sulfenylmethanimine.
Insights into the Photodecomposition of Azidomethyl Methyl Sulfide: A S2/S1 Conical Intersection on Nitrene Potential Energy Surfaces Leading to the Formation of S-Methyl-N-sulfenylmethanimine.
The journal of physical chemistry. A (2020-02-14)
Manuel Algarra, Juan Soto, Luis Pinto da Silva, M Soledad Pino-González, J Enrique Rodríguez-Borges, Joelle Mascetti, Fabien Borget, Adel Reisi-Vanani, Rafael Luque
RÉSUMÉ
UV photodecomposition of azidomethyl methyl sulfide (AMMS) yields a transient S-methylthiaziridine which rapidly evolves to S-methyl-N-sulfenylmethanimine at 10 K. This species was detected by infrared matrix isolation spectroscopy. The mechanism of the photoreaction of AMMS has been investigated by a combined approach, using low-temperature matrix isolation FTIR spectroscopy in conjunction with two theoretical methods, namely, complete active space self-consistent field and multiconfigurational second-order perturbation. The key step of the reaction is governed by a S2/S1 conical intersection localized in the neighborhood of the singlet nitrene minimum which is formed in the first reaction step of the photolysis, that is, N2 elimination from AMMS. Full assignment of the observed infrared spectra of AMMS has been carried out based on comparison with density functional theory and second-order perturbation Møller-Plesset methods.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Chloromethyl methyl sulfide, 95%