MilliporeSigma
ROCK2 Confers Acquired Gemcitabine Resistance in Pancreatic Cancer Cells by Upregulating Transcription Factor ZEB1.
Cancers (2019-12-01)
Yang Zhou, Yunjiang Zhou, Keke Wang, Tao Li, Minda Zhang, Yunjia Yang, Rui Wang, Rong Hu
RÉSUMÉ
Resistance to chemotherapy is a major clinical challenge in the treatment of pancreatic ductal adenocarcinoma (PDAC). Here, we provide evidence that Rho associated coiled-coil containing protein kinase 2 (ROCK2) maintains gemcitabine resistance in gemcitabine resistant pancreatic cancer cells (GR cells). Pharmacological inhibition or gene silencing of ROCK2 markedly sensitized GR cells to gemcitabine by suppressing the expression of zinc-finger-enhancer binding protein 1 (ZEB1). Mechanically, ROCK2-induced sp1 phosphorylation at Thr-453 enhanced the ability of sp1 binding to ZEB1 promoter regions in a p38-dependent manner. Moreover, transcriptional activation of ZEB1 facilitated GR cells to repair gemcitabine-mediated DNA damage via ATM/p-CHK1 signaling pathway. Our findings demonstrate the essential role of ROCK2 in EMT-induced gemcitabine resistance in pancreatic cancer cells and provide strong evidence for the clinical application of fasudil, a ROCK2 inhibitor, in gemcitabine-refractory PDAC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Mithramycin A from Streptomyces plicatus, ≥90% (HPLC)