MilliporeSigma
Exchange protein directly activated by cAMP (Epac) protects against airway inflammation and airway remodeling in asthmatic mice.
Respiratory research (2019-12-20)
Yi-Fei Chen, Ge Huang, Yi-Min Wang, Ming Cheng, Fang-Fang Zhu, Jin-Nan Zhong, Ya-Dong Gao
RÉSUMÉ
β2 receptor agonists induce airway smooth muscle relaxation by increasing intracellular cAMP production. PKA is the traditional downstream signaling pathway of cAMP. Exchange protein directly activated by cAMP (Epac) was identified as another important signaling molecule of cAMP recently. The role of Epac in asthmatic airway inflammation and airway remodeling is unclear. We established OVA-sensitized and -challenged acute and chronic asthma mice models to explore the expression of Epac at first. Then, airway inflammation and airway hyperresponsiveness in acute asthma mice model and airway remodeling in chronic asthma mice model were observed respectively after treatment with Epac-selective cAMP analogue 8-pCPT-2'-O-Me-cAMP (8pCPT) and Epac inhibitor ESI-09. Next, the effects of 8pCPT and ESI-09 on the proliferation and apoptosis of in vitro cultured mouse airway smooth muscle cells (ASMCs) were detected with CCK-8 assays and Annexin-V staining. Lastly, the effects of 8pCPT and ESI-09 on store-operated Ca2+ entry (SOCE) of ASMCs were examined by confocal Ca2+ fluorescence measurement. We found that in lung tissues of acute and chronic asthma mice models, both mRNA and protein expression of Epac1 and Epac2, two isoforms of Epac, were lower than that of control mice. In acute asthma mice model, the airway inflammatory cell infiltration, Th2 cytokines secretion and airway hyperresponsiveness were significantly attenuated by 8pCPT and aggravated by ESI-09. In chronic asthma mice model, 8pCPT decreased airway inflammatory cell infiltration and airway remodeling indexes such as collagen deposition and airway smooth muscle cell proliferation, while ESI-09 increased airway inflammation and airway remodeling. In vitro cultured mice ASMCs, 8pCPT dose-dependently inhibited, whereas ESI-09 promoted ASMCs proliferation. Interestingly, 8pCPT promoted the apoptosis of ASMCs, whereas ESI-09 had no effect on ASMCs apoptosis. Lastly, confocal Ca2+ fluorescence examination found that 8pCPT could inhibit SOCE in ASMCs at 100 μM, and ESI-09 promoted SOCE of ASMCs at 10 μM and 100 μM. In addition, the promoting effect of ESI-09 on ASMCs proliferation was inhibited by store-operated Ca2+ channel blocker, SKF-96365. Our results suggest that Epac has a protecting effect on asthmatic airway inflammation and airway remodeling, and Epac reduces ASMCs proliferation by inhibiting SOCE in part.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
8-(4-Chlorophenylthio)-2′-O-methyladenosine 3′,5′-cyclic monophosphate monosodium hydrate, ≥98% (HPLC)