MilliporeSigma
Activation of caspases and mitochondria in FTY720-mediated apoptosis in human T cell line Jurkat.
International immunopharmacology (2001-10-19)
M Fujino, X K Li, L Guo, T Amano, S Suzuki
RÉSUMÉ
FTY720, a novel immunosuppressive drug originally derived from a metabolite from Isaria sinclairii, is known to induce apoptosis in lymphocytes. In this study, we investigated the involvement of caspases and mitochondria in FTY720-mediated apoptosis using Jurkat cells, a human T cell line. Our results indicated that FTY720-induced activation of caspases 2, 3, 6, 8, 9 and 10, whereas caspases 1 and 5 were not activated. We also observed in the FTY720-treated cells a loss of mitochondrial membrane potential, a release of cytochrome c into cytosol and an exposed phosphatidylserine (PS) at the outer surface of the cell membrane. Pretreatment with a peptide inhibitor, benzyloxycarbonyl-Asp-CH2COC-2, 6-dichlorobenzene (Z-Asp-CH2-DCB), prevented apoptosis and externalization of phosphatidylserine, whereas the inhibitor did not prevent the mitochondrial events. This suggests that caspases may play a role downstream of the mitochondrial pathway. Therefore, caspase cascade in FTY720-treated cells may be initiated by activation of mitochondria.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Caspase Substrate