MilliporeSigma
Structure-function aspects in the nitric oxide synthases.
Annual review of pharmacology and toxicology (1997-01-01)
D J Stuehr
RÉSUMÉ
Research on the biological roles of nitric oxide has revealed that it functions as an important signal and effector molecule in a variety of physiologic and pathologic settings. In animals, nitric oxide is synthesized enzymatically from L-arginine through the actions of the nitric oxide synthases (NOSs). The three known NOS isoforms are all dimeric, bi-domain enzymes that contain iron protoporphyrin IX, flavin adenine dinucleotide, flavin mononucleotide, and tetrahydrobiopterin as bound prosthetic groups. This chapter summarizes information regarding the structure-function aspects of the NOSs, which includes composition of the domains, the protein residues and regions involved in prosthetic group binding, catalytic properties of the domains, the relationship between dimeric structure and prosthetic group binding and function, and factors that control assembly of NOS in cells. A general model for NOS structure and assembly is presented.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Nitric Oxide Synthase Detection System, Fluorimetric, sufficient for 200 reactions
Le tarif et la disponibilité ne sont pas disponibles actuellement.