MilliporeSigma
Long Noncoding RNA Moderates MicroRNA Activity to Maintain Self-Renewal in Embryonic Stem Cells.
Stem cell reports (2017-06-06)
Keriayn N Smith, Joshua Starmer, Sarah C Miller, Praveen Sethupathy, Terry Magnuson
RÉSUMÉ
Of the thousands of long noncoding RNAs expressed in embryonic stem cells (ESCs), few have known roles and fewer have been functionally implicated in the regulation of self-renewal and pluripotency, or the reprogramming of somatic cells to the pluripotent state. In ESCs, Cyrano is a stably expressed long intergenic noncoding RNA with no previously assigned role. We demonstrate that Cyrano contributes to ESC maintenance, as its depletion results in the loss of hallmarks of self-renewal. Delineation of Cyrano's network through transcriptomics revealed widespread effects on signaling pathways and gene expression networks that contribute to ESC maintenance. Cyrano shares unique sequence complementarity with the differentiation-associated microRNA, mir-7, and mir-7 overexpression reduces expression of a key self-renewal factor to a similar extent as Cyrano knockdown. This suggests that Cyrano functions to restrain the action of mir-7. Altogether, we provide a view into the multifaceted function of Cyrano in ESC maintenance.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Ago2 Antibody, clone 9E8.2, ascites fluid, clone 9E8.2, Upstate®