MilliporeSigma
  • Central Treatment of Ketone Body in Rainbow Trout Alters Liver Metabolism Without Apparently Altering the Regulation of Food Intake.
Central Treatment of Ketone Body in Rainbow Trout Alters Liver Metabolism Without Apparently Altering the Regulation of Food Intake.
Frontiers in physiology (2019-10-18)
Sara Comesaña, Cristina Velasco, Marta Conde-Sieira, Cristina Otero-Rodiño, Jesús M Míguez, José L Soengas
RÉSUMÉ
We hypothesize that the presence in fish brain of a ketone body (KB) like β-hydroxybutyrate (BHB) alters energy homeostasis through effects on food intake and peripheral energy metabolism. Using rainbow trout (Oncorhynchus mykiss) as a model, we intracerebroventricularly (ICV) administered 1 μl 100 g-1 body mass of saline solution alone (control) or containing 0.5 μmol of BHB. In a fist set of experiments, BHB did not affect food intake 6 and 24 h after treatment. In a second set of experiments, we evaluated 6 h after ICV BHB treatment changes in parameters putatively related to food intake control in brain areas (hypothalamus and hindbrain) involved in nutrient sensing and changes in energy metabolism in liver. The absence of changes in food intake might relate to the absence of major changes in the cascade of events from the detection of KB through ketone-sensing mechanisms, changes in transcription factors, and changes in the mRNA abundance of neuropeptides regulating food intake. This response is different than that of mammals. In contrast, central administration of BHB induced changes in liver energy metabolism suggesting a decreased use of glucose and probably an enhanced use of amino acid and lipid. These responses in liver are different to those of mammals under similar treatments but comparable to those occurring in fish under food deprivation conditions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-mTOR antibody produced in rabbit, 1.0-1.5 mg/mL, affinity isolated antibody, buffered aqueous solution