MilliporeSigma
Molecular and cellular activities of vitamin E analogues.
Mini reviews in medicinal chemistry (2007-05-17)
Jean-Marc Zingg
RÉSUMÉ
Natural vitamin E comprises 8 different analogues, the alpha-, beta-, gamma-, and delta-tocopherols and the alpha-, beta-, gamma-, and delta-tocotrienols. However, only alpha-tocopherol is selectively enriched by the liver; the other vitamin E analogues and also excess alpha-tocopherol are converted to several metabolites and eliminated. Recently, a novel phosphorylated form of tocopherol, alpha-tocopheryl phosphate, was shown to occur naturally in animal and human tissues as well as in foods. Several synthetic vitamin E derivatives have been synthesized that are either converted by esterases to the natural form, or exert novel or vitamin E related biological activities. During the last years, specific cellular effects for each individual vitamin E analogue have been described that are the consequence of modulating signal transduction and gene expression. These effects possibly reflect specific interactions of each of the vitamin E analogues with enzymes, structural proteins, lipids and transcription factors. In this review, the different natural vitamin E analogues and synthetic derivatives are compiled in relation to their major molecular and cellular activities.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(±)-α-Tocopherol phosphate disodium salt, water-soluble α-tocopherol analog