MilliporeSigma
An integrated genome-wide multi-omics analysis of gene expression dynamics in the preimplantation mouse embryo.
Scientific reports (2019-09-19)
Steffen Israel, Mathias Ernst, Olympia E Psathaki, Hannes C A Drexler, Ellen Casser, Yutaka Suzuki, Wojciech Makalowski, Michele Boiani, Georg Fuellen, Leila Taher
RÉSUMÉ
Early mouse embryos have an atypical translational machinery that consists of cytoplasmic lattices and is poorly competent for translation. Hence, the impact of transcriptomic changes on the operational level of proteins is predicted to be relatively modest. To investigate this, we performed liquid chromatography-tandem mass spectrometry and mRNA sequencing at seven developmental stages, from the mature oocyte to the blastocyst, and independently validated our data by immunofluorescence and qPCR. We detected and quantified 6,550 proteins and 20,535 protein-coding transcripts. In contrast to the transcriptome - where changes occur early, mostly at the 2-cell stage - our data indicate that the most substantial changes in the proteome take place towards later stages, between the morula and blastocyst. We also found little to no concordance between the changes in protein and transcript levels, especially for early stages, but observed that the concordance increased towards the morula and blastocyst, as did the number of free ribosomes. These results are consistent with the cytoplasmic lattice-to-free ribosome transition being a key mediator of developmental regulation. Finally, we show how these data can be used to appraise the strengths and limitations of mRNA-based studies of pre-implantation development and expand on the list of known developmental markers.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-TOP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DNAJB11 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution