MilliporeSigma
Acylated Ghrelin and The Regulation of Lipid Metabolism in The Intestine.
Scientific reports (2019-12-01)
N Auclair, N Patey, L Melbouci, Y Ou, L Magri-Tomaz, A Sané, C Garofalo, E Levy, D H St-Pierre
RÉSUMÉ
Acylated ghrelin (AG) is a gastrointestinal (GI) peptide mainly secreted by the stomach that promotes cytosolic lipid droplets (CLD) hypertrophy in adipose tissues and liver. However, the role of AG in the regulation of lipid metabolism in the intestine remains unexplored. This study aimed at determining whether AG influences CLD production and chylomicron (CM) secretion in the intestine. The effects of AG and oleic acid on CLD accumulation and CM secretion were first investigated in cultured Caco-2/15 enterocytes. Intestinal lipid metabolism was also studied in Syrian Golden Hamsters submitted to conventional (CD) or Western (WD) diets for 8 weeks and continuously administered with AG or physiological saline for the ultimate 2 weeks. In cultured Caco-2/15 enterocytes, CLD accumulation influenced CM secretion while AG reduced fatty acid uptake. In WD hamsters, continuous AG treatment amplified chylomicron output while reducing postprandial CLD accumulation in the intestine. The present study supports the intimate relationship between CLD accumulation and CM secretion in the intestine and it underlines the importance of further characterizing the mechanisms through which AG exerts its effects on lipid metabolism in the intestine.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-Apolipoprotein B Antibody, clone B1, clone B1, Chemicon®, from mouse