MilliporeSigma
Highly Enantioselective Rhodium-Catalyzed Addition of Arylboroxines to Simple Aryl Ketones: Efficient Synthesis of Escitalopram.
Angewandte Chemie (International ed. in English) (2016-03-05)
Linwei Huang, Jinbin Zhu, Guangjun Jiao, Zheng Wang, Xingxin Yu, Wei-Ping Deng, Wenjun Tang
RÉSUMÉ
Highly enantioselective additions of arylboroxines to simple aryl ketones have been achieved for the first time with a Rh/(R,R,R,R)-WingPhos catalyst, thus providing a range of chiral diaryl alkyl carbinols with excellent ee values and yields. (R,R,R,R)-WingPhos has been proven to be crucial for the high reactivity and enantioselectivity. The method has enabled a new, concise, and enantioselective synthesis of the antidepressant drug escitalopram.