MilliporeSigma
Knockdown of TRIM28 inhibits PDGF-BB-induced vascular smooth muscle cell proliferation and migration.
Chemico-biological interactions (2019-07-28)
Hongtao Liu, Hongwei Chen, Xia Deng, Yudong Peng, Qiutang Zeng, Zongren Song, Wenping He, Le Zhang, Ting Xiao, Gan Gao, Bailin Li
RÉSUMÉ
Atherosclerosis is a common type of cardiovascular disease (CVD), remaining one of the leading causes of global death. Tripartite motif-containing 28 (TRIM28) is a member of TRIM family that has been found to be involved in atherosclerosis. However, the role of TRIM28 in atherosclerosis remains unknown. This study aimed to investigate the effects of TRIM28 on the phenotypic switching of human aortic smooth muscle cells (HASMCs), which is considered as a fundamental event during the development of atherosclerosis. The results showed that TRIM28 was highly expressed in human atherosclerotic tissues, as well in cultured HASMCs stimulated by platelet-derived growth factor subunit B homodimer (PDGF-BB). Knockdown of TRIM28 by transfection with siRNA targeting TRIM28 (si-TRIM28) significantly suppressed the PDGF-BB-induced cell proliferation and migration of HASMCs. Besides, knockdown of TRIM28 inhibited the expressions of matrix metalloproteinase (MMP)-2 and MMP-9. The VSMC markers including α-smooth muscle actin (α-SMA), calponin and SM22α were upregulated in TRIM28 knocked down HASMCs. Furthermore, knockdown of TRIM28 blocked PDGF-BB-induced NF-κB activation in HASMCs. Collectively, knockdown of TRIM28 prevented PDGF-BB-induced phenotypic switching of HASMCs, which might be mediated by the regulation of NF-κB signaling pathway.