MilliporeSigma
  • FCPR16, a novel phosphodiesterase 4 inhibitor, produces an antidepressant-like effect in mice exposed to chronic unpredictable mild stress.
FCPR16, a novel phosphodiesterase 4 inhibitor, produces an antidepressant-like effect in mice exposed to chronic unpredictable mild stress.
Progress in neuro-psychopharmacology & biological psychiatry (2018-11-06)
Qiuping Zhong, Hui Yu, Chang Huang, Jiahong Zhong, Haitao Wang, Jiangping Xu, Yufang Cheng
RÉSUMÉ
The canonical phosphodiesterase 4 (PDE4) inhibitors produce antidepressant-like effects in a variety of animal models. However, severe side effects, particularly vomiting and nausea, limit their clinical application. FCPR16 is a novel PDE4 inhibitor with less vomiting potential. However, whether it will exert an antidepressant-like effect remains unclear. Here, we aimed to evaluate the effect of FCPR16 in mice subjected to chronic unpredictable mild stress (CUMS). Our results showed that FCPR16 produced antidepressant-like effects in multiple behavioral tests, including a forced swimming test, tail suspension test, sucrose preference test and novelty suppression feeding test. Simultaneously, data indicated that FCPR16 enhanced the levels of several proteins, including cAMP, brain derived neurotrophic factor, exchange protein directly activated by cAMP 2 (EPAC-2), synapsin1, postsynaptic density protein 95, phosphorylated cAMP response element binding protein and extracellular regulated protein kinases 1/2, which were downregulated by CUMS in both the cerebral cortex and hippocampus. The number of DCX+ cells in the hippocampus of CUMS mice was increased after FCPR16 treatment. Moreover, treatment with FCPR16 resulted in decreased expression of pro-inflammatory cytokines (TNF-α, IL-6, and IL-1β) and increased expression of anti-inflammatory cytokines (IL-10) in mice challenged with CUMS. Consistently, the mRNA levels of microglial M1 markers (iNOS and TNF-α) were downregulated, while M2 markers (Arginase 1 and CD206) were upregulated in CUMS-exposed mice after FCPR16 treatment. Immunofluorescence analysis showed that FCPR16 inhibited the activation of microglial cells and increased the number of CD206+ in CUMS-exposed mice. Collectively, these results suggested that FCPR16 is a potential compound with effects against depressive-like behaviors, and the antidepressant-like effect of FCPR16 is possibly mediated through activation of the cAMP-mediated signaling pathways and inhibition of neuroinflammation in both the cerebral cortex and hippocampus.