MilliporeSigma
Poly(butylene terephthalate) based novel achiral stationary phase investigated under supercritical fluid chromatography conditions.
Journal of chromatography. A (2018-03-28)
Kanji Nagai, Tohru Shibata, Satoshi Shinkura, Atsushi Ohnishi
RÉSUMÉ
Poly(butylene terephthalate) based novel stationary phase (SP), composed of planar aromatic phenyl group together with ester group monomer units, was designed for supercritical fluid chromatography (SFC) use. As expected from its structure, this phase shows planarity recognition of isomeric aromatics and closely similar compounds. Interestingly, for most analytes, the retention behavior of this SP is significantly distinct from that of the 2-ethylpyridine based SPs which is among the most well-known SFC dedicated phases. Although the poly(butylene terephthalate) is coated on silica gel, the performance of the column did not change by using extended range modifiers such as THF, dichloromethane or ethyl acetate and column robustness was confirmed by cycle durability testing.