MilliporeSigma
Identification of substituted 5-membered heterocyclic compounds as potential anti-leukemic agents.
European journal of medicinal chemistry (2019-01-07)
Taotao Ling, Julie Maier, Sourav Das, Amit Budhraja, Rachel Bassett, Malia B Potts, Anang Shelat, Zoran Rankovic, Joseph T Opferman, Fatima Rivas
RÉSUMÉ
Although pediatric leukemia is generally treatable, certain leukemic subtypes face poor prognosis in the clinic suggesting new selective therapeutic agents are needed. Thus, to identify selective apoptosis inducers, a small-molecule library screening approach was conducted using an isogenic leukemic murine p185+ B-ALL cell line pair (BCR-ABL-WT and the BAX/BAK deficient BCR-ABL-DKO). Gratifyingly, the investigation revealed several compounds featuring substituted aromatic five-membered-ring heterocycles with significant activity against murine and human leukemic cellular models. The identified compounds represent potentially novel antileukemic molecular scaffolds exemplified by compounds 1, 2 and 7, which demonstrated EC50 values in the nanomolar and low micromolar range against various leukemia subtypes (SUP-B15, KOPN-8, NALM-06, UoC-B1 cellular models) and pro-apoptotic properties in solid tumor cell models (MDA-MB-231, SUM149) with ample therapeutic index in normal cells. Herein, we highlight compounds 1, 2 and 7 which promote cell death mediated by caspase 3/7 induction. Our study establishes a strategic platform for the development of potent and selective anti-leukemic agents.