MilliporeSigma
  • High-selective HDAC6 inhibitor promotes HDAC6 degradation following autophagy modulation and enhanced antitumor immunity in glioblastoma.
High-selective HDAC6 inhibitor promotes HDAC6 degradation following autophagy modulation and enhanced antitumor immunity in glioblastoma.
Biochemical pharmacology (2019-03-20)
Jia-Rong Liu, Chao-Wu Yu, Pei-Yun Hung, Ling-Wei Hsin, Ji-Wang Chern
RÉSUMÉ
Glioblastoma is the most fatal type of primary brain cancer, and current treatments for glioblastoma are insufficient. HDAC6 is overexpressed in glioblastoma, and siRNA-mediated knockdown of HDAC6 inhibits glioma cell proliferation. Herein, we report a high-selective HDAC6 inhibitor, J22352, which has PROTAC (proteolysis-targeting chimeras)-like property resulted in both p62 accumulation and proteasomal degradation, leading to proteolysis of aberrantly overexpressed HDAC6 in glioblastoma. The consequences of decreased HDAC6 expression in response to J22352 decreased cell migration, increased autophagic cancer cell death and significant tumor growth inhibition. Notably, J22352 reduced the immunosuppressive activity of PD-L1, leading to the restoration of host anti-tumor activity. These results demonstrate that J22352 promotes HDAC6 degradation and induces anticancer effects by inhibiting autophagy and eliciting the antitumor immune response in glioblastoma. Therefore, this highly selective HDAC6 inhibitor can be considered a potential therapeutic for the treatment of glioblastoma and other cancers.