MilliporeSigma
  • Identification and Characterization of Synthetic Viability with ERCC1 Deficiency in Response to Interstrand Crosslinks in Lung Cancer.
Identification and Characterization of Synthetic Viability with ERCC1 Deficiency in Response to Interstrand Crosslinks in Lung Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-12-13)
Joshua R Heyza, Wen Lei, Donovan Watza, Hao Zhang, Wei Chen, Jessica B Back, Ann G Schwartz, Gerold Bepler, Steve M Patrick
RÉSUMÉ
ERCC1/XPF is a DNA endonuclease with variable expression in primary tumor specimens, and has been investigated as a predictive biomarker for efficacy of platinum-based chemotherapy. The failure of clinical trials utilizing ERCC1 expression to predict response to platinum-based chemotherapy suggests additional mechanisms underlying the basic biology of ERCC1 in the response to interstrand crosslinks (ICLs) remain unknown. We aimed to characterize a panel of ERCC1 knockout (Δ) cell lines, where we identified a synthetic viable phenotype in response to ICLs with ERCC1 deficiency. We utilized the CRISPR-Cas9 system to create a panel of ERCC1Δ lung cancer cell lines which we characterized. We observe that loss of ERCC1 hypersensitizes cells to cisplatin when wild-type (WT) p53 is retained, whereas there is only modest sensitivity in cell lines that are p53mutant/null. In addition, when p53 is disrupted by CRISPR-Cas9 (p53*) in ERCC1Δ/p53WT cells, there is reduced apoptosis and increased viability after platinum treatment. These results were recapitulated in 2 patient data sets utilizing p53 mutation analysis and ERCC1 expression to assess overall survival. We also show that kinetics of ICL-repair (ICL-R) differ between ERCC1Δ/p53WT and ERCC1Δ/p53* cells. Finally, we provide evidence that cisplatin tolerance in the context of ERCC1 deficiency relies on DNA-PKcs and BRCA1 function. Our findings implicate p53 as a potential confounding variable in clinical assessments of ERCC1 as a platinum biomarker via promoting an environment in which error-prone mechanisms of ICL-R may be able to partially compensate for loss of ERCC1.See related commentary by Friboulet et al., p. 2369.