MilliporeSigma
Nonylphenol acts on prostate adenocarcinoma cells via estrogen molecular pathways.
Ecotoxicology and environmental safety (2019-05-21)
Maurizio Forte, Mariana Di Lorenzo, Giuseppina Iachetta, Damiano Gustavo Mita, Vincenza Laforgia, Maria De Falco
RÉSUMÉ
Estrogens play a role in the patho-physiology of the prostate. In the present work we studied the effects of nonylphenol (NP), a xenoestrogen, on human adenocarcinoma prostate cells (LNCaP). In order to understand molecular and cellular involvement, we observed the effects on cell cycle and we investigated the expression and the cellular localization of estrogen receptors and gene expression of cyclin D1, ki-67, c-myc, IL-8, IL-1β. We performed the same experiments with 17β-estradiol (E2), the most abundant estrogen circulating in nonpregnant humans in order to compare these two different substances. We demonstrated the ability of 1 × 10-10 M NP to induce proliferation of LNCaP, S-phase progression, increase of ERα expression and its translocation from the cytoplasm to the nucleus. Moreover, we observed an up-regulation of key target genes involved in cell cycle and inflammation process. Particularly, after NP treatment, IL-8 and IL-1β mRNA levels are increased more than 50% indicating a major NP involvement in inflammation processes than E2. These data suggest the proliferative effects of NP on prostate adenocarcinoma cells and highlight some aspects of molecular pathways involved in prostate responses to NP.