MilliporeSigma
Zinc-binding to the cytoplasmic PAS domain regulates the essential WalK histidine kinase of Staphylococcus aureus.
Nature communications (2019-07-13)
Ian R Monk, Nausad Shaikh, Stephanie L Begg, Mike Gajdiss, Liam K R Sharkey, Jean Y H Lee, Sacha J Pidot, Torsten Seemann, Michael Kuiper, Brit Winnen, Rikki Hvorup, Brett M Collins, Gabriele Bierbaum, Saumya R Udagedara, Jacqueline R Morey, Neha Pulyani, Benjamin P Howden, Megan J Maher, Christopher A McDevitt, Glenn F King, Timothy P Stinear
RÉSUMÉ
WalKR (YycFG) is the only essential two-component regulator in the human pathogen Staphylococcus aureus. WalKR regulates peptidoglycan synthesis, but this function alone does not explain its essentiality. Here, to further understand WalKR function, we investigate a suppressor mutant that arose when WalKR activity was impaired; a histidine to tyrosine substitution (H271Y) in the cytoplasmic Per-Arnt-Sim (PASCYT) domain of the histidine kinase WalK. Introducing the WalKH271Y mutation into wild-type S. aureus activates the WalKR regulon. Structural analyses of the WalK PASCYT domain reveal a metal-binding site, in which a zinc ion (Zn2+) is tetrahedrally-coordinated by four amino acids including H271. The WalKH271Y mutation abrogates metal binding, increasing WalK kinase activity and WalR phosphorylation. Thus, Zn2+-binding negatively regulates WalKR. Promoter-reporter experiments using S. aureus confirm Zn2+ sensing by this system. Identification of a metal ligand recognized by the WalKR system broadens our understanding of this critical S. aureus regulon.