MilliporeSigma
The biochemical activity of soil contaminated with fungicides.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2019-01-11)
Małgorzata Baćmaga, Jadwiga Wyszkowska, Jan Kucharski
RÉSUMÉ
Excess fungicides can pose a serious threat to the soil environment. Fungicides can lower the microbiological and biochemical activity of soil and lead to yield declines. Soils contaminated with fungicides have to be remediated to maintain the optimal function of soil ecosystems. This study evaluates the effect of neutralizing substances on soil enzymatic activity and the yield of Triticum aestivum L. in soil contaminated with fungicides. Sandy loam (Eutric Cambisols) with pHKCl 7.0 was contaminated with an aqueous solution of Amistar 250 SC and Falcon 460 EC in the following doses: 0 (soil without fungicide - treated as a control), RD (dose recommended by the manufacturer) and 300 × RD (dose 300-fold higher than the recommended dose). Soil was supplemented with bentonite and basalt meal at a dose of 10 g kg-1 DM of soil (dry mass of soil). The fungicide dose recommended by the manufacturer did not induce changes in soil enzymatic activity or the yield of T. aestivum L. Our findings indicate that the tested fungicides can be safely applied to protect crops against fungal pathogens. However, when applied at the dose of 300 × RD, the tested fungicides strongly inhibited soil enzymatic activity and disrupted the growth and development of spring wheat. Soil supplementation with bentonite and basalt meal improved the yield of T. aestivum L., and bentonite was more effective in reducing fungicide stress. The analyzed substances were not highly effective in restoring biochemical homeostasis in soil.