MilliporeSigma
Silymarin protects against radiocontrast-induced nephropathy in mice.
Life sciences (2019-05-03)
Verônica de Souza Santos, Beatriz Peters, Larissa Zambom Côco, Gisele Maziero Alves, Arícia Leone Evangelista Monteiro de Assis, Breno Valentim Nogueira, Silvana Santos Meyrelles, Marcella Leite Porto, Elisardo Corral Vasquez, Bianca Prandi Campagnaro, Thiago Melo Costa Pereira
RÉSUMÉ
Silymarin, an extract from Silybum marianum (milk thistle) containing a standardized mixture of flavonolignans that ameliorates some types of liver disease and, more recently, kidney damage, could be used for the ROS-scavenging effect of these antioxidants. Furthermore, contrast-induced nephropathy (CIN) is an iatrogenic impairment of renal function in patients subjected to angiographic procedures for which there is not yet a successful preventative treatment. Recent evidence has shown that this event is related to tubular/vascular injury activated mainly by oxidative stress. However, whether this bioavailable and pharmacologically safe extract protects against CIN is not clear. We proposed to evaluate the possible protective role of the antioxidant silymarin in an experimental model of CIN. Adult male Swiss mice were separated into 6 groups and pretreated orally with silymarin (50, 200 and 300 mg/kg), N-acetylcysteine (200 mg/kg) or vehicle for 5 days before the CIN and control groups. Renal function was analyzed by plasma creatinine, urea and cystatin C levels. Additionally, blood reactive oxygen species (ROS) were evaluated using ROS bioavailability, protein oxidation and DNA damage. Renal oxidative damage was evaluated using apoptosis/cell viability assays and histological analysis. We showed that silymarin preserved renal function and decreased systemic and renal oxidative damage (antigenotoxic and antiapoptotic properties, respectively) in a dose-dependent manner and was superior to conventional treatment with N-acetylcysteine. Histologically, silymarin treatment also had beneficial effects on renal glomerular and tubular injuries. Therefore, silymarin prophylaxis may be an interesting strategy for the prevention of CIN.