MilliporeSigma
  • Protein-engineered molecules carrying GAD65 epitopes and targeting CD35 selectively down-modulate disease-associated human B lymphocytes.
Protein-engineered molecules carrying GAD65 epitopes and targeting CD35 selectively down-modulate disease-associated human B lymphocytes.
Clinical and experimental immunology (2019-04-23)
I K Manoylov, G V Boneva, I A Doytchinova, N M Mihaylova, A I Tchorbanov
RÉSUMÉ
Type 1 diabetes mellitus is an autoimmune metabolic disorder characterized by chronic hyperglycemia, the presence of autoreactive T and B cells and autoantibodies against self-antigens. A membrane-bound enzyme on the pancreatic beta-cells, glutamic acid decarboxylase 65 (GAD65), is one of the main autoantigens in type 1 diabetes. Autoantibodies against GAD65 are potentially involved in beta-cell destruction and decline of pancreatic functions. The human complement receptor type 1 (CD35) on B and T lymphocytes has a suppressive activity on these cells. We hypothesized that it may be possible to eliminate GAD65-specific B cells from type 1 diabetes patients by using chimeric molecules, containing an anti-CD35 antibody, coupled to peptides resembling GAD65 B/T epitopes. These molecules are expected to selectively bind the anti-GAD65 specific B cells by the co-cross-linking of the immunoglobulin receptor and CD35 and to deliver a suppressive signal. Two synthetic peptides derived from GAD65 protein (GAD65 epitopes) and anti-CD35 monoclonal antibody were used for the construction of two chimeras. The immunomodulatory activity of the engineered antibodies was tested in vitro using peripheral blood mononuclear cells (PBMCs) from type 1 diabetes patients. A reduction in the number of anti-GAD65 IgG antibody-secreting plasma cells and increased percentage of apoptotic B lymphocytes was observed after treatment of these PBMCs with the engineered antibodies. The constructed chimeric molecules are able to selectively modulate the activity of GAD65-specific B lymphocytes and the production of anti-GAD65 IgG autoantibodies by co-cross-linking of the inhibitory CD35 and the B cell antigen receptor (BCR). This treatment presents a possible way to alter the autoimmune nature of these cells.