MilliporeSigma
  • Evaluation of the enantioselectivity of capillary electrokinetic chromatography using ethanediamine-bonded poly (glycidyl methacrylate) microspheres as the pseudostationary phases.
Evaluation of the enantioselectivity of capillary electrokinetic chromatography using ethanediamine-bonded poly (glycidyl methacrylate) microspheres as the pseudostationary phases.
Chirality (2019-01-05)
Xiaodong Sun, Tao Yu, Guangfu Xu, Yingxiang Du, Jiaquan Chen, Xiaoqi Li
RÉSUMÉ
In this work, a new capillary electrokinetic chromatography (EKC) approach using ethanediamine-bonded poly (glycidyl methacrylate) (Ami-PGMA) microspheres as pseudostationary phases (PSPs) for enantioseparation with a polysaccharide, chondroitin sulfate E (CSE), as the chiral selector. The CSE@Ami-PGMA EKC system was applied to enantioseparate basic drugs, and distinct improved separations of tested enantiomers were obtained while comparing with the single CSE system (the resolution increased from 0.41 to 1.26 for nefopam, from 1.24 to 2.15 for laudanosine, and from 0.92 to 2.36 for amlodipine). The Ami-PGMA microspheres were fully characterized by scanning electron microscopy (SEM) and Fourier Transform Infrared (FT-IR) spectroscopy, and the results showed Ami-PGMA microspheres were uniform and spherical in size (1 μm). Several principal parameters were systematically investigated, and the optimal chiral separations were obtained with Tris/H3 PO4 (20 mM, pH 2.4, and 3.4 for NEF) containing 2.5% (w/v) CSE and 20-μg Ami-PGMA microspheres in 20°C. Subsequently, the concentrations of Ami-PGMA microspheres and CSE were proved to be the dominant factors for the separation in the CSE@Ami-PGMA EKC system by Statistical Product and Service Solutions (SPSS).