MilliporeSigma
  • Targeted analysis of microbial-generated phenolic acid metabolites derived from grape flavanols by gas chromatography-triple quadrupole mass spectrometry.
Targeted analysis of microbial-generated phenolic acid metabolites derived from grape flavanols by gas chromatography-triple quadrupole mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2018-07-23)
Eileen Carry, Danyue Zhao, Ilaria Mogno, Jeremiah Faith, Lap Ho, Tom Villani, Harna Patel, Giulio M Pasinetti, James E Simon, Qingli Wu
RÉSUMÉ
Grape-derived products contain a wide array of bioactive phenolic compounds which are of significant interest to consumers and researchers for their multiple health benefits. The majority of bioavailable grape polyphenols, including the most abundant flavan-3-ols, i.e. (+)-catechin and (-)-epicatechin, undergo extensive microbial metabolism in the gut, forming metabolites that can be highly bioavailable and bioactive. To gain a better understanding in microbial metabolism of grape polyphenols and to identify bioactive metabolites, advanced analytical methods are needed to accurately quantitate microbial-derived metabolites, particularly at trace levels, in addition to their precursors. This work describes the development and validation of a high-throughput, sensitive and reproducible GC-QqQ/MS method operated under MRM mode that allowed the identification and quantification of 16 phenolic acid metabolites, along with (+)-catechin and (-)-epicatechin, in flavanol-enriched broth samples anaerobically fermented with human intestinal bacteria. Excellent sensitivity was achieved with low limits of detection and low limits of quantification in the range of 0.24-6.18 ng/mL and 0.480-12.37 ng/mL, respectively. With the exception of hippuric acid, recoveries of most analytes were greater than 85%. The percent accuracies for almost all analytes were within ±23% and precision results were all below 18%. Application of the developed method to in vitro samples fermented with different human gut microbiota revealed distinct variations in the extent of flavanol catabolism, as well as production of bioactive phenolic acid metabolites. These results support that intestinal microbiota have a significant impact on the production of flavanol metabolites. The successful application of the established method demonstrates its applicability and robustness for analysis of grape flavanols and their microbial metabolites in biological samples.