MilliporeSigma
The applicability of furfuryl-gelatin as a novel bioink for tissue engineering applications.
Journal of biomedical materials research. Part B, Applied biomaterials (2018-04-16)
Shweta AnilKumar, Shane C Allen, Nishat Tasnim, Tahmina Akter, Shinhye Park, Alok Kumar, Munmun Chattopadhyay, Yoshihiro Ito, Laura J Suggs, Binata Joddar
RÉSUMÉ
Three-dimensional bioprinting is an innovative technique in tissue engineering, to create layer-by-layer structures, required for mimicking body tissues. However, synthetic bioinks do not generally possess high printability and biocompatibility at the same time. So, there is an urgent need for naturally derived bioinks that can exhibit such optimized properties. We used furfuryl-gelatin as a novel, visible-light crosslinkable bioink for fabricating cell-laden structures with high viability. Hyaluronic acid was added as a viscosity enhancer and either Rose Bengal or Riboflavin was used as a visible-light crosslinker. Crosslinking was done by exposing the printed structure for 2.5 min to visible light and confirmed using Fourier transform infrared spectroscopy and rheometry. Scanning electron microscopy revealed a highly porous networked structure. Three different cell types were successfully bioprinted within these constructs. Mouse mesenchymal stem cells printed within monolayer and bilayer sheets showed viability, network formation and proliferation (∼5.33 times) within 72 h of culture. C2C12 and STO cells were used to print a double layered structure, which showed evidence of the viability of both cells and heterocellular clusters within the construct. This furfuryl-gelatin based bioink can be used for tissue engineering of complex tissues and help in understanding how cellular crosstalk happens in vivo during normal or diseased pathology. © 2018 Wiley Periodicals, Inc. J Biomed Mater Res Part B: Appl Biomater, 107B: 314-323, 2019.